Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndrg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013865.2 | NP_038893.1 | 16 | 2629 | ||
NM_180956.1 | NP_851287.1 | 17 | 2668 | |||
GenBank mRNA | AB033922.1 | - | 16 | 2543 | ||
AK004689.1 | - | 16 | 2575 | |||
AK037487.1 | - | 16 | 2555 | |||
AK154927.1 | - | 16 | 2596 | |||
AK211130.1 | - | 1 | 357 | |||
AK217093.1 | - | 1 | 385 | |||
AV032677.1 | - | 1 | 191 | |||
BC018504.1 | - | 17 | 2602 | 4500970 |