Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp385a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013866.2 | NP_038894.2 | 8 | 1695 | ||
GenBank mRNA | AF118566.1 | - | 8 | 1689 | ||
AK137748.1 | - | 8 | 1702 | |||
BC017644.1 | - | 8 | 1752 | 4504518 |