Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapk12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013871.3 | NP_038899.1 | 11 | 1205 | ||
GenBank mRNA | AK011286.1 | - | 10 | 1245 | ||
BC002134.1 | - | 1 | 746 | |||
BC021640.1 | - | 11 | 1123 | 3979667 | ||
Y13439.1 | - | 11 | 1141 |