Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klf15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK177169.1 | - | 29 |
Gene Symbol: Mapk12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013871.3 | NP_038899.1 | 11, 12 | 1262 | ||
GenBank mRNA | AA855742.1 | - | 1, 2 | 51 | ||
AK011286.1 | - | 10, 11 | 1302 | |||
BC002134.1 | - | 1, 2 | 803 | |||
BC021640.1 | - | 11, 12 | 1180 | 3979667 | ||
Y13439.1 | - | 11, 12 | 1198 |