Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sult4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013873.3 | NP_038901.3 | 7 | 1631 | ||
GenBank mRNA | AK032195.1 | - | 7 | 1517 | ||
AK046368.1 | - | 7 | 1510 | |||
AK159034.1 | - | 7 | 1523 | |||
BC051132.1 | - | 7 | 1504 | 6531493 | ||
BC054757.1 | - | 7 | 1631 | 6413084 |