Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013874.2 | NP_038902.1 | 5, 6 | 603 | ||
GenBank mRNA | AK142419.1 | - | 5, 6 | 582 | ||
BC052348.1 | - | 5, 6 | 632 | 6587002 | ||
U48238.1 | - | 5, 6 | 602 |