Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013874.2 | NP_038902.1 | 12 | 1780 | ||
GenBank mRNA | U48238.1 | - | 12 | 1776 |