Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtse1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168672.1 | NP_001162143.1 | 4 | 422 | ||
NM_013882.2 | NP_038910.1 | 3 | 392 | |||
GenBank mRNA | AJ222580.1 | - | 3 | 362 | ||
AK012870.1 | - | 4 | 423 | |||
AK081582.1 | - | 4 | 385 | |||
BC027213.1 | - | 3 | 393 | 4039841 |