Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scmh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159630.1 | NP_001153102.1 | 10 | 1362 | ||
NM_013883.2 | NP_038911.1 | 10 | 1362 | |||
GenBank mRNA | AB030906.1 | - | 10 | 1261 | ||
AB221680.1 | - | 6 | 903 | |||
AB221681.1 | - | 6 | 903 | |||
BC025478.1 | - | 1 | 1604 | |||
BC034667.1 | - | 8 | 1072 | 5356322 |