Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdgfrp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013886.4 | NP_038914.2 | 4 | 866 | ||
GenBank mRNA | AB029493.1 | - | 4 | 429 | ||
AK014849.1 | - | 4 | 690 | |||
AK019487.1 | - | 4 | 529 | |||
AK044858.1 | - | 4 | 691 | |||
AK045542.1 | - | 4 | 866 | |||
AK051401.1 | - | 4 | 788 | |||
AK162418.1 | - | 4 | 509 | |||
AK162967.1 | - | 4 | 430 | |||
BC055734.1 | - | 4 | 453 | 6810355 |