Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Opn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013887.2 | NP_038915.1 | 9 | 1932 | ||
GenBank mRNA | AF147789.1 | - | 9 | 1932 | ||
BC139827.1 | - | 9 | 1873 | 40142922 | ||
EU303118.1 | - | 9 | 1748 |
Set Membership: |
G-protein coupled receptor |