Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timm13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013895.4 | NP_038923.1 | 3 | 545 | ||
GenBank mRNA | AF144702.1 | - | 3 | 547 | ||
AK003054.1 | - | 3 | 351 | |||
AK009041.1 | - | 3 | 339 | |||
AK009152.1 | - | 3 | 355 | |||
AK013230.1 | - | 3 | 343 | |||
AK088984.1 | - | 3 | 355 | |||
BC011436.1 | - | 3 | 358 | 2812494 |