Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timm13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013895.4 | NP_038923.1 | 3 | 885 | ||
GenBank mRNA | AK009041.1 | - | 3 | 679 | ||
AK013230.1 | - | 3 | 683 | |||
AK213641.1 | - | 1 | 295 |