Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Heyl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013905.3 | NP_038933.2 | 2, 3 | 457 | ||
GenBank mRNA | AB093590.1 | - | 2, 3 | 149 | ||
AF172288.1 | - | 2, 3 | 457 | |||
AJ271868.2 | - | 2, 3 | 457 | |||
AK004697.2 | - | 2, 3 | 160 | |||
AK028596.1 | - | 2, 3 | 196 | |||
AK080822.1 | - | 2, 3 | 161 | |||
BC130263.1 | - | 2, 3 | 165 | 40142873 |