Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxw5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013908.4 | NP_038936.1 | 6 | 1186 | ||
GenBank mRNA | AF176520.1 | - | 6 | 1147 | ||
AK019715.2 | - | 6 | 1187 | |||
AK154911.1 | - | 5 | 2125 | |||
AK187599.1 | - | 1 | 149 | |||
BC010776.1 | - | 6 | 1113 | 4211592 |