Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxl12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002846.2 | NP_001002846.1 | 3 | 1542 | ||
NM_001286529.1 | NP_001273458.1 | 3 | 1164 | |||
NM_001286530.1 | NP_001273459.1 | 4 | 1268 | |||
NM_013911.3 | NP_038939.2 | 3 | 1572 | |||
GenBank mRNA | AF176525.1 | - | 3 | 1453 | ||
AK011081.1 | - | 3 | 1527 | |||
AK012811.1 | - | 3 | 1065 | |||
AK029429.1 | - | 1 | 507 | |||
AK030910.1 | - | 3 | 1542 | |||
AK031262.1 | - | 2 | 3628 | |||
AK137282.1 | - | 4 | 3239 | |||
BC005699.1 | - | 1 | 1058 | 3597990 | ||
BC054471.1 | - | 3 | 1572 | 4039251 |