Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnf4g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013920.2 | NP_038948.1 | 7 | 1003 | ||
GenBank mRNA | AJ242626.1 | - | 7 | 955 | ||
AK143816.1 | - | 7 | 1004 | |||
BC125519.1 | - | 7 | 825 | 40130034 |