Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Schip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113420.1 | NP_001106891.1 | 2 | 274 | ||
NM_001113421.1 | NP_001106892.1 | 3 | 958 | |||
NM_001282044.1 | NP_001268973.1 | 3 | 941 | |||
NM_001282045.1 | NP_001268974.1 | 3 | 799 | |||
NM_013928.5 | NP_038956.2 | 2 | 364 | |||
GenBank mRNA | AF145716.1 | - | 2 | 470 | ||
AK021361.1 | - | 2 | 225 | |||
AK083464.1 | - | 3 | 784 | |||
AK146930.1 | - | 3 | 915 | |||
AK168005.1 | - | 3 | 868 | |||
BC060529.1 | - | 2 | 298 | |||
BC094348.1 | - | 3 | 867 | 6414097 | ||
BY243167.1 | - | 2 | 365 | |||
EU163407.1 | - | 3 | 958 | |||
EU163408.1 | - | 2 | 274 | |||
KM233715.1 | - | 2 | 220 | |||
KM233716.1 | - | 2 | 225 |
Gene Symbol: Gm21949 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113419.2 | NP_001106890.1 | 6 | 1136 | ||
GenBank mRNA | KJ941154.1 | - | 5 | 440 |