Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Schip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113420.1 | NP_001106891.1 | 5 | 693 | ||
NM_001113421.1 | NP_001106892.1 | 6 | 1377 | |||
NM_001282044.1 | NP_001268973.1 | 6 | 1360 | |||
NM_001282045.1 | NP_001268974.1 | 6 | 1218 | |||
NM_013928.5 | NP_038956.2 | 5 | 783 | |||
GenBank mRNA | AF145716.1 | - | 5 | 889 | ||
AK021361.1 | - | 5 | 644 | |||
AK083464.1 | - | 6 | 1203 | |||
AK146930.1 | - | 6 | 1334 | |||
AK168005.1 | - | 6 | 1287 | |||
BC060529.1 | - | 5 | 717 | |||
BC094348.1 | - | 6 | 1286 | 6414097 | ||
EU163407.1 | - | 6 | 1377 | |||
EU163408.1 | - | 5 | 693 | |||
KM233715.1 | - | 5 | 639 | |||
KM233716.1 | - | 5 | 644 |
Gene Symbol: Gm21949 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113419.2 | NP_001106890.1 | 9 | 1555 | ||
GenBank mRNA | KJ941154.1 | - | 8 | 859 |