Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aass | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013930.4 | NP_038958.2 | 3, 4 | 516 | ||
GenBank mRNA | AJ224761.1 | - | 2, 3 | 407 | ||
AK044323.1 | - | 3, 4 | 497 | |||
AK098141.1 | - | 3, 4 | 457 | |||
AK136216.1 | - | 3, 4 | 465 | |||
AK149400.1 | - | 3, 4 | 441 | |||
BC005420.1 | - | 3, 4 | 502 | 3600654 |