Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aass | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013930.4 | NP_038958.2 | 14 | 1540 | ||
GenBank mRNA | AJ224761.1 | - | 13 | 1431 | ||
AK044323.1 | - | 14 | 1521 | |||
AK098141.1 | - | 14 | 1481 | |||
AK136216.1 | - | 14 | 1489 | |||
AK149400.1 | - | 14 | 1465 | |||
BC005420.1 | - | 14 | 1526 | 3600654 |