Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013932.4 | NP_038960.2 | 9 | 1046 | ||
GenBank mRNA | AF142630.3 | - | 9 | 977 | ||
AK050693.1 | - | 9 | 1041 | |||
AK078340.1 | - | 9 | 1005 | |||
BC024852.1 | - | 9 | 990 | 5358404 | ||
BC061130.1 | - | 9 | 1001 | 6774579 |