Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shroom3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015756.2 | NP_056571.2 | 1 | 378 | ||
GenBank mRNA | AF199421.1 | - | 1 | 378 | ||
AK018481.1 | - | 1022 | ||||
BC145113.1 | - | 1 | 188 | 9053661 |