Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDE10A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130690.2 | NP_001124162.1 | 22 | 3104 | ||
NM_006661.3 | NP_006652.1 | 23 | 3232 | |||
XM_006715321.3 | XP_006715384.1 | 22 | 4429 | |||
XM_011535387.2 | XP_011533689.2 | 24 | 5224 | |||
XM_011535388.2 | XP_011533690.1 | 23 | 4557 | |||
XM_011535393.2 | XP_011533695.1 | 22 | 3032 | |||
XM_017010194.1 | XP_016865683.1 | 24 | 5228 | |||
XM_017010195.1 | XP_016865684.1 | 22 | 2977 | |||
XM_017010196.1 | XP_016865685.1 | 24 | 4675 | |||
GenBank mRNA | AB020593.1 | - | 23 | 3236 | ||
AF127479.1 | - | 22 | 3053 | |||
AF127480.1 | - | 22 | 3073 | |||
AK021956.1 | - | 6 | 1329 | |||
KU232567.1 | - | 22 | 3475 |