Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ero1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015774.3 | NP_056589.1 | 13 | 1185 | ||
GenBank mRNA | AK009667.1 | - | 3 | 410 | ||
AK142595.1 | - | 13 | 1176 | |||
BC025102.1 | - | 13 | 1137 | 2654098 | ||
CT010195.1 | - | 13 | 1103 | |||
CT010259.1 | - | 13 | 1103 |