Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmprss2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015775.2 | NP_056590.2 | 5 | 567 | ||
GenBank mRNA | AF113596.1 | - | 5 | 437 | ||
AF199362.1 | - | 5 | 448 | |||
AF243500.1 | - | 5 | 437 | |||
AK141350.1 | - | 5 | 542 | |||
AK146047.1 | - | 5 | 451 | |||
AK161173.1 | - | 5 | 561 | |||
BC038393.1 | - | 5 | 406 | 2648878 | ||
BC054348.1 | - | 5 | 567 | 2648889 |