Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nap1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146707.1 | NP_001140179.1 | 14 | 1930 | ||
GenBank mRNA | AK012250.1 | - | 11 | 938 | ||
AK050375.1 | - | 14 | 1932 | |||
AK190527.1 | - | 4 | 257 | |||
BC076591.1 | - | 14 | 1478 | 30618066 | ||
X61449.1 | - | 12 | 1099 |