Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snrpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001046637.1 | NP_001040102.1 | 5 | 861 | ||
NM_001285825.1 | NP_001272754.1 | 6 | 935 | |||
NM_015782.3 | NP_056597.3 | 6 | 938 | |||
GenBank mRNA | AK013886.1 | - | 6 | 912 | ||
AK088550.1 | - | 6 | 794 | |||
AK209909.1 | - | 1 | 34 | |||
BC003229.1 | - | 6 | 838 | 3483391 | ||
BC094006.1 | - | 6 | 798 | 4983854 | ||
BC096648.1 | - | 6 | 792 | 1397390 | ||
DV650448.1 | - | 4 | 554 | |||
L15447.1 | - | 6 | 734 |