Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289507.1 | NP_001276436.1 | 18 | 2695 | ||
NM_001289509.1 | NP_001276438.1 | 20 | 2714 | |||
NM_001289511.1 | NP_001276440.1 | 19 | 2572 | |||
NM_015799.4 | NP_056614.3 | 20 | 2654 | |||
GenBank mRNA | AF207741.1 | - | 18 | 2529 | ||
AF222895.1 | - | 19 | 2572 | |||
AK004848.2 | - | 20 | 2630 | |||
AK004965.1 | - | 20 | 2685 | |||
AK155526.1 | - | 19 | 2791 | |||
AK165320.1 | - | 18 | 2522 | |||
BB736789.1 | - | 1 | 77 | |||
BC013654.1 | - | 18 | 2695 | 4196597 |