Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168320.1 | NP_001161792.1 | 8 | 1324 | ||
NM_006675.4 | NP_006666.1 | 9 | 1391 | |||
XM_011520912.2 | XP_011519214.1 | 10 | 1508 | |||
XM_011520913.2 | XP_011519215.1 | 7 | 1250 | |||
GenBank mRNA | AF089749.1 | - | 9 | 1334 | ||
AK027366.1 | - | 9 | 1367 | |||
AK223179.1 | - | 9 | 1363 | |||
AY523992.1 | - | 1 | 154 | |||
BC071881.1 | - | 8 | 1306 | 6256950 |