Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015810.2 | NP_056625.2 | 4 | 919 | ||
GenBank mRNA | AF177202.1 | - | 4 | 824 | ||
AK041591.1 | - | 4 | 891 | |||
AK042777.1 | - | 4 | 730 | |||
AK169977.1 | - | 4 | 1161 | |||
BC119131.1 | - | 4 | 848 | 8733880 | ||
BC144810.1 | - | 4 | 723 |