Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dkk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015814.2 | NP_056629.1 | 3 | 389 | ||
GenBank mRNA | AF177400.1 | - | 3 | 376 | ||
AJ243964.1 | - | 3 | 392 | |||
AK004853.1 | - | 3 | 389 | |||
AK013054.2 | - | 2 | 327 | |||
AK013622.1 | - | 2 | 301 | |||
AK199048.1 | - | 1 | 34 | |||
BC005519.1 | - | 2 | 88 | |||
BC046304.1 | - | 2 | 304 | 6493718 | ||
BC050934.1 | - | 2 | 286 | 6812621 |