Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppap2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005452.1 | - | 3 | 448 | ||
AK049581.1 | - | 3 | 491 | |||
AK179023.1 | - | 2 | 252 | |||
AK184662.1 | - | 1 | 83 | |||
AK208995.1 | - | 2 | 197 | |||
AK214880.1 | - | 2 | 220 | |||
BC010332.1 | - | 3 | 469 | 2654393 | ||
BC016635.1 | - | 3 | 419 | |||
BC085104.1 | - | 3 | 407 | |||
BQ571983.1 | - | 2 | 294 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AF123611.1 | - | 483 | Chr. - - [-] on Build |