Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ME3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014811.1 | NP_001014811.1 | 14 | 1969 | ||
NM_001161586.1 | NP_001155058.1 | 15 | 2042 | |||
NM_006680.2 | NP_006671.2 | 15 | 1817 | |||
XM_005273717.1 | XP_005273774.1 | 15 | 1918 | |||
XM_017017136.1 | XP_016872625.1 | 15 | 2800 | |||
XM_017017137.1 | XP_016872626.1 | 11 | 1342 | |||
XM_017017138.1 | XP_016872627.1 | 16 | 1926 | |||
GenBank mRNA | AK313695.1 | - | 15 | 1892 | ||
BC022472.2 | - | 14 | 1969 | 4812045 | ||
BQ882896.1 | - | 6 | 703 | |||
X79440.1 | - | 14 | 1747 |