Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Keap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110305.1 | NP_001103775.1 | 2 | 742 | ||
NM_001110306.1 | NP_001103776.1 | 2 | 627 | |||
NM_001110307.1 | NP_001103777.1 | 2 | 459 | |||
NM_016679.4 | NP_057888.1 | 1 | 1778 | |||
GenBank mRNA | AB020063.1 | - | 2 | 231 | ||
AK004738.1 | - | 2 | 459 | |||
AK039549.1 | - | 2 | 397 | |||
AK046178.1 | - | 2 | 343 | |||
AK076234.1 | - | 1 | 1264 | |||
AK129061.1 | - | 1 | 213 | |||
AK142272.1 | - | 1 | 1271 | |||
AK150437.1 | - | 2 | 363 | |||
AK150485.1 | - | 2 | 230 | |||
AK152142.1 | - | 2 | 363 | |||
AK154430.1 | - | 2 | 274 | |||
AK155507.1 | - | 2 | 274 | |||
AK159858.1 | - | 2 | 228 | |||
BC006729.1 | - | 2 | 240 | |||
BC023945.1 | - | 2 | 492 | |||
BC055732.1 | - | 1 | 216 | 6404252 | ||
CJ179527.1 | - | 2 | 375 |