Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Keap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110305.1 | NP_001103775.1 | 4 | 2076 | ||
NM_001110306.1 | NP_001103776.1 | 4 | 1961 | |||
NM_001110307.1 | NP_001103777.1 | 4 | 1793 | |||
NM_016679.4 | NP_057888.1 | 3 | 3112 | |||
GenBank mRNA | AB020063.1 | - | 4 | 1565 | ||
AK004738.1 | - | 4 | 1793 | |||
AK039549.1 | - | 4 | 1732 | |||
AK046178.1 | - | 4 | 1677 | |||
AK076234.1 | - | 3 | 2598 | |||
AK129061.1 | - | 3 | 1547 | |||
AK136319.1 | - | 2 | 797 | |||
AK150437.1 | - | 4 | 1697 | |||
AK150485.1 | - | 4 | 1564 | |||
AK152142.1 | - | 4 | 1697 | |||
AK154430.1 | - | 4 | 1608 | |||
AK155507.1 | - | 4 | 1608 | |||
AK159858.1 | - | 4 | 1562 | |||
BC055732.1 | - | 3 | 1550 | 6404252 |