Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Keap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110305.1 | NP_001103775.1 | 6 | 2794 | ||
NM_001110306.1 | NP_001103776.1 | 6 | 2679 | |||
NM_001110307.1 | NP_001103777.1 | 6 | 2511 | |||
NM_016679.4 | NP_057888.1 | 5 | 3830 | |||
GenBank mRNA | AK004738.1 | - | 6 | 2511 | ||
AK039549.1 | - | 6 | 2450 | |||
AK046178.1 | - | 6 | 2395 | |||
AK076234.1 | - | 5 | 3316 | |||
AK129061.1 | - | 5 | 2265 | |||
AK136319.1 | - | 4 | 1515 | |||
AK150437.1 | - | 6 | 2415 | |||
AK150485.1 | - | 6 | 2282 | |||
AK152142.1 | - | 6 | 2415 | |||
AK159858.1 | - | 6 | 2279 |