Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chek2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016681.3 | NP_057890.1 | 5 | 713 | ||
GenBank mRNA | AF086905.1 | - | 4 | 674 | ||
AK044913.1 | - | 5 | 714 | |||
BC056617.1 | - | 5 | 737 | 4977960 |