Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016698.2 | NP_057907.2 | 3 | 907 | ||
GenBank mRNA | AB026621.1 | - | 3 | 906 | ||
AK004579.1 | - | 3 | 901 | |||
AK034953.1 | - | 3 | 906 | |||
AK080426.1 | - | 1 | 790 | |||
AK129106.1 | - | 3 | 895 | |||
AK146728.1 | - | 3 | 907 | |||
AK149972.1 | - | 3 | 908 | |||
AK156502.1 | - | 3 | 904 | |||
AK165617.1 | - | 3 | 907 | |||
AK168523.1 | - | 3 | 902 | |||
AK172065.1 | - | 3 | 905 | |||
AK213843.1 | - | 1 | 56 | |||
BC010342.1 | - | 3 | 616 | 3602969 |