Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmgn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016710.2 | NP_057919.2 | 5 | 250 | ||
GenBank mRNA | AB018374.2 | - | 5 | 205 | ||
AF213454.1 | - | 5 | 230 | |||
AK013748.1 | - | 4 | 119 | |||
AK131996.1 | - | 5 | 220 | |||
AK146755.1 | - | 5 | 226 | |||
BC021626.1 | - | 5 | 202 | 4219308 | ||
BC083087.1 | - | 5 | 212 | 6518798 | ||
BY279928.1 | - | 5 | 275 |