Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmod2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038710.1 | NP_001033799.1 | 6 | 954 | ||
NM_016711.3 | NP_057920.2 | 6 | 890 | |||
GenBank mRNA | AF177170.1 | - | 6 | 839 | ||
AF237629.1 | - | 7 | 798 | |||
AK017072.1 | - | 6 | 938 | |||
AK076562.1 | - | 6 | 904 | |||
AK079077.1 | - | 6 | 734 | |||
AK147581.1 | - | 6 | 886 | |||
AK164178.1 | - | 6 | 736 | |||
BC052227.1 | - | 6 | 954 | |||
BC061124.1 | - | 6 | 720 | 6531518 |