Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ninj2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016718.2 | NP_057927.1 | 3 | 473 | ||
GenBank mRNA | AF205634.1 | - | 3 | 503 | ||
AK138486.1 | - | 3 | 463 | |||
AK163748.1 | - | 3 | 463 | |||
BC116206.1 | - | 3 | 458 | 40090174 |