Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grb14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016719.1 | NP_057928.1 | 10 | 1281 | ||
GenBank mRNA | AF155647.1 | - | 10 | 1281 | ||
AK147041.1 | - | 10 | 1290 | |||
BC021820.1 | - | 3 | 167 | |||
BC138581.1 | - | 10 | 1187 | 8861601 | ||
BC145913.1 | - | 10 | 1187 | 40131216 |