Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nub1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF534114.1 | - | 2, 3 | 296 | ||
AK018166.1 | - | 3, 4 | 347 | |||
AK086301.1 | - | 3, 4 | 506 | |||
AK134581.1 | - | 3, 4 | 350 | |||
AK153400.1 | - | 3, 4 | 334 | |||
BC005640.1 | - | 3, 4 | 335 | |||
BC018178.1 | - | 3, 4 | 321 | |||
BC037451.1 | - | 2, 3 | 325 | 4018648 | ||
U27462.1 | - | 3, 4 | 543 |