Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccnc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290422.1 | NP_001277351.1 | 13 | 1664 | ||
NM_016746.4 | NP_058026.2 | 12 | 1570 | |||
GenBank mRNA | AK037957.1 | - | 13 | 1665 | ||
BC100396.1 | - | 12 | 1544 | |||
BC120649.1 | - | 12 | 1497 | 40129572 | ||
BC120677.1 | - | 12 | 1497 | 40129600 | ||
U62638.1 | - | 12 | 1546 |