Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177778.2 | NP_001171249.1 | 7 | 1353 | ||
NM_001290402.1 | NP_001277331.1 | 6 | 1299 | |||
NM_016747.4 | NP_058027.1 | 7 | 1353 | |||
GenBank mRNA | AK122474.1 | - | 7 | 1352 | ||
BC060713.1 | - | 5 | 1331 | |||
BC094368.1 | - | 6 | 1294 | 6842105 | ||
D87117.1 | - | 7 | 1327 |