Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177778.2 | NP_001171249.1 | 20 | 3480 | ||
NM_001177779.1 | NP_001171250.1 | 14 | 2477 | |||
NM_001177780.1 | NP_001171251.1 | 12 | 1869 | |||
NM_001290402.1 | NP_001277331.1 | 19 | 3426 | |||
NM_016747.4 | NP_058027.1 | 21 | 3522 | |||
GenBank mRNA | AK122474.1 | - | 20 | 3479 | ||
AK145815.1 | - | 14 | 2232 | |||
AK148464.1 | - | 12 | 1849 | |||
AK159655.1 | - | 12 | 1857 | |||
AK167365.1 | - | 12 | 1856 | |||
BC060713.1 | - | 19 | 3554 | |||
BC094368.1 | - | 19 | 3421 | 6842105 | ||
D87117.1 | - | 21 | 3496 |