Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mybph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016749.2 | NP_058029.2 | 11 | 1636 | ||
GenBank mRNA | AK029449.1 | - | 4 | 1035 | ||
BC116684.1 | - | 11 | 1603 | 40106746 | ||
BC118050.1 | - | 11 | 1606 | 40106744 | ||
U68267.1 | - | 11 | 1630 |