Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016758.3 | NP_058038.2 | 14 | 1650 | ||
GenBank mRNA | AK002891.1 | - | 9 | 1139 | ||
AK154944.1 | - | 13 | 1471 | |||
BC030321.1 | - | 14 | 1622 | 5374531 | ||
U85055.1 | - | 14 | 1490 |