Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016758.3 | NP_058038.2 | 15 | 2105 | ||
GenBank mRNA | AK002891.1 | - | 10 | 1594 | ||
AK154944.1 | - | 14 | 1926 | |||
BC030321.1 | - | 15 | 2077 | 5374531 |